missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Bulk, Linerless Polyester Cable Tags for M611, M610, BMP61, M710 and BMP71
Bulk, Linerless Polyester Cable Tags for use with Brady™ Printers M611, M610, BMP61, M710 and BMP71.
£203.00 - £390.00
Specifications
Type | Tag |
---|---|
Certifications/Compliance | RoHS Directive 2011/65/EU |
Material | Polyester |
Finish | Gloss |
For Use With (Application) | Data and Telecommunications Labeling, Wire and Cable Labeling |
Product Code | Brand | Color | Height (Metric) | Length (English) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Height (Metric) | Length (English) | Width (Metric) | Price | Quantity & Availability | ||
17898064
|
Brady™
312142 |
Black | 60 mm | 2.362 in. | 10 mm |
£390.00
Pack of 500 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17848044
|
Brady™
312149 |
Black | 75 mm | 2.953 in. | 10 mm |
£245.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17867904
|
Brady™
312156 |
Black | 75 mm | 2.953 in. | 15 mm |
£245.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17898074
|
Brady™
312163 |
Black | 75 mm | 2.953 in. | 25 mm |
£302.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17808074
|
Brady™
312146 |
Silver | 60 mm | 2.362 in. | 10 mm |
£390.00
Pack of 500 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17857894
|
Brady™
312153 |
Silver | 75 mm | 2.953 in. | 10 mm |
£245.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17887944
|
Brady™
312160 |
Silver | 75 mm | 2.953 in. | 15 mm |
£245.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17858004
|
Brady™
312167 |
Silver | 75 mm | 2.953 in. | 25 mm |
£302.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17817984
|
Brady™
312145 |
Red | 60 mm | 2.362 in. | 10 mm |
£390.00
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17897894
|
Brady™
312152 |
Red | 75 mm | 2.953 in. | 10 mm |
£245.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17857904
|
Brady™
312159 |
Red | 75 mm | 2.953 in. | 15 mm |
£245.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17878094
|
Brady™
312166 |
Red | 75 mm | 2.953 in. | 25 mm |
£302.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17817894
|
Brady™
312148 |
Yellow | 60 mm | 2.362 in. | 10 mm |
£321.00
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17877894
|
Brady™
312155 |
Yellow | 75 mm | 2.953 in. | 10 mm |
£203.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17847914
|
Brady™
312162 |
Yellow | 75 mm | 2.953 in. | 15 mm |
£203.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17857924
|
Brady™
312169 |
Yellow | 75 mm | 2.953 in. | 25 mm |
£252.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17897844
|
Brady™
312144 |
Green | 60 mm | 2.362 in. | 10 mm |
£390.00
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17807984
|
Brady™
312151 |
Green | 75 mm | 2.953 in. | 10 mm |
£245.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17827914
|
Brady™
312158 |
Green | 75 mm | 2.953 in. | 15 mm |
£245.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17827854
|
Brady™
312165 |
Green | 75 mm | 2.953 in. | 25 mm |
£302.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17877884
|
Brady™
312143 |
Blue | 60 mm | 2.362 in. | 10 mm |
£390.00
Pack of 500 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17858074
|
Brady™
312150 |
Blue | 75 mm | 2.953 in. | 10 mm |
£245.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17827904
|
Brady™
312157 |
Blue | 75 mm | 2.953 in. | 15 mm |
£245.00
Pack of 250 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17888074
|
Brady™
312164 |
Blue | 75 mm | 2.953 in. | 25 mm |
£302.00
Pack of 250 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
Description
- B-7598 polyester cable identification tag has been developed to keep cables identified in outdoor environments
- Offers up to 10 year outdoor legibility and durability
- Available in multiple colors
Specifications
Tag | |
Polyester | |
Data and Telecommunications Labeling, Wire and Cable Labeling | |
0.2 mm | |
-40°F to +212°F | |
5 x 2 mm |
RoHS Directive 2011/65/EU | |
Gloss | |
Rectangle | |
-40°C to +100°C | |
0.19 x 0.07 in. | |
Brady™ Printers M610, M710, M611, BMP61, BMP71 |