missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bulk tubes
GREENER_CHOICE

Product Code. p-8750002 Shop All Fisherbrand Products
Click to view available options
Product Type:
Non-reinforced Tubes
Reinforced Tubes
Unit Size:
1 pack

Product Code. 15545809

Only null left
Add to basket
Edge
Add to basket

For use with Bead Mill 4 and Bead Mill 24 homogenizers. Also fits most other commercially available Bead Mills.

  • RNase/DNase free
  • Processing range: 300μL to 1.8mL
  • Beads not included
TRUSTED_SUSTAINABILITY

Specifications

Product Type Reinforced Tubes
Capacity (Metric) 2 mL
Color White/Opaque
Dnase Free Yes
For Use With (Equipment) Bead Mill 4 and 24 Homogenizers
Includes Screw Caps and O-rings
Packaging Quantity 500/Pk.
RNase Free Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.