Learn More
Fisherbrand™ Bunsen Burner Neoprene Tubing
Product Code. p-8011940 Shop All Fisherbrand Products
Click to view available options
Overall Length (Metric):
10 m
2 m
450 mm
600 mm
750 mm
900 mm
Unit Size:
10m
2m
Each
Product Code. 12330768
Only null left
Add to basket
Edge
Add to basket
Suitable for use with coal and natural (methane) gas burners.
With reinforced ends for fitting to Bunsen burner and gas tap
Specifications
Material Neoprene
Overall Length (Metric) 10 m
For Use With Bunsen Burner