missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Dry-blend Buffered Packs
Our quality control ensures that every pack of dry-blend buffers yields consistent solutions without weighing or pH adjustment. Simply empty contents of one foil envelope pack into a beaker, add appropriate solvents, and stir to dissolve.
£75.50 - £185.00
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
10627524
|
Thermo Scientific™
28372 |
Phosphate Buffered Saline (PBS) | 40 Packs |
£163.00
Pack of 40 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10494404
|
Thermo Scientific™
28390 |
MES Buffered Saline | 10 Packs |
£133.00
Pack of 10 |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10331414
|
Thermo Scientific™
28382 |
Carbonate-Bicarbonate Buffer Packs | 40 Packs |
£152.00
Pack of 40 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10137833
|
Thermo Scientific™
28376 |
Tris Buffered Saline Packs | 40 Packs |
£167.00
Pack of 40 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10628694
|
Thermo Scientific™
28384 |
Borate Buffer | 40 Packs |
£163.00
Pack of 40 |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10501294
|
Thermo Scientific™
28380 |
Tris-Glycine Buffer | 40 Packs |
£185.00
Pack of 40 |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10350884
|
Thermo Scientific™
28379 |
Tris Buffered Saline Packs | 10 Packs |
£75.50
Pack of 10 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10197443
|
Thermo Scientific™
28374 |
DPBS | 40 Packs |
£166.00
Pack of 40 |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Our dry-blend buffer packs are easy to use. Simply empty contents of one foil envelope pack into a beaker, add ultrapure water, and stir to dissolve. No need to make and store large volumes of stock solution in advance of daily needs.
Benefits of ready-to-use buffered packs include:• Convenient—no weighing, no pH adjustment, no need to stock individual components
• Consistent quality—our quality control ensures that every pack yields the same, consistent buffer
• Long shelf life—stocking and storage as dry packs eliminates concerns about long-term stability of stock solutions
BupH™ Phosphate-Buffered Saline Packs are pouches of dry-blend powder that make sodium phosphate buffered saline (PBS) for use in crosslinking and labeling reactions requiring amine-free conditions. Each BupH Dry-Blend pack makes ready-to-use PBS with 0.1 M sodium phosphate, 0.15 M NaCl buffer at pH 7.2 when dissolved in 500 mL of water.
BupH MES-Buffered Saline Packs are pouches of dry-blended powder that make optimized amine-free and carboxyl-free buffer. When dissolved in 500 mL of water, the buffer contains 0.1 M 2-(N-morpholino) ethanesulfonic acid (MES), 0.9% sodium chloride at pH 4.7 for carbodiimide (EDC) crosslinking and conjugation reactions.
BupH Carbonate-Bicarbonate Buffer Packs are pouches of dry-blended powder that are each sufficient to make 500 mL of alkaline (pH 9.4) sodium-carbonate (carb-bicarb) buffer. This buffer is used for coating or binding capture-antibodies and other proteins onto polystyrene microplates for ELISA and other plate assays. When dissolved in 500 mL of water, each pack makes 0.2 M carbonate-bicarbonate at pH 9.4.
BupH Tris-Buffered Saline Packs are pouches of dry-blend powder that make Tris-NaCl buffered saline (TBS) for diluents and wash buffers in ELISA, western blotting, and general immunoassay procedures. Each pack makes ready-to-use TBS with 25 mM Tris, 0.15 M NaCl at pH 7.2 when dissolved in 500 mL of water.
BupH Borate Buffer Packs are pouches of dry-blended powder that are each sufficient to make borate buffer for use in protein modification procedures requiring amine-free conditions. When dissolved in 500 mL of water, each pack makes 50 mM borate at pH 8.5.
BupH Tris-Glycine Transfer Buffer Packs are pouches of dry-blend powder that make 500 mL of standard transfer buffer for wet or semi-dry electrophoretic protein transfer from gel to blotting membrane. Each pack makes 25 mM Tris, 192 mM glycine at pH 8 when dissolved in 400 mL of water and 100 mL methanol.
BupH Modified Dulbecco's PBS Packs are pouches of dry-blend powder that make physiological sodium- and potassium-phosphate buffered saline (D-PBS) for ELISA and western blot diluents and wash buffers. Each pack makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl modified Dulbecco's PBS buffer at pH 7.4 when dissolved in 500 mL of water.
Specifications
BupH™ | |
Powder |