missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Technotrate Digital Burette
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
Beschreibung
- Developed in compliance with EN ISO 9001 and it is CE certified
- Adequately allows dosage volumes of up to 50 ml with 0.01 ml increases to be covered and the dosage velocity can be selected from fast mode to drop-by-drop mode
- Simple calibration
- Permanent calibration refers to water at 20°C
- When using liquids with a different viscosity/density, Technotrate can easily and quickly be recalibrated by pressing the dedicated button
- Practical and quick preselection of the titration volumes
- Titration volume preselection is made particularly simple by pressing only the dedicated DOS button, looking at the display, and with just a few turns of the handle the burette cylinder is filled
- The special memory button (MEM) helps you avoid complex calculations
- Easy and safe
- The components that come into contact with the reagents are made of high resistance plastic.
- Tested one by one at the end of production, has quality certification and an individual serial number
Spezifikation
Spezifikation
| Accuracy | ±0.2% |
| Description | Technotrate Digital Burette |
| Volume (Metric) | 50 mL |
| Includes | Auto off funtion after 3 minutes of non-use |
| Increments | 0.01 mL |
| Model | Technotrate |
| Precision | ±0.1% |
| Temperature (Metric) | 10°C to 40°C |
| Voltage | 100/240 V |
| Frequency | 50/60 Hz |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur