Learn More
Lenz Laborglasintrumente Burette Stopcock, Straight, with PTFE Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02700501
Product Code. 10479742
  • £32.35 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Facetted tip
  • Stopcock: NS 12.5
  • Bore: 1.5 mm
  • NS PTFE plug
  • Screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Specifications
Borosilicate Glass (3.3)
NS 12.5