Learn More
Thermo Scientific™ Burkholderia cepacia Sel Agar (USP 60) 500g
Compliance to the USP 60 regulations for B. cepacia complex made simple
Brand: Thermo Scientific™ R452642
Description
Burkholderia cepacia medium (USP 60) is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in products, especially those for inhalation use of non-sterile acqueous preparations for oral, oromucosal, cutaneous or nasal use.
Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.
- Compliant formulation according to USP 60
- Quality tested to meet USP 60 specifically for Pharma customers
- Part of a complete workflow which include DCM and QCO

Specifications
Burkholderia cepacia Selective agar (USP 60) | |
Agar | |
500 g |
500g Pot | |
Pharmaceutical testing of non-sterile products used for nasal inhalation | |
Lab use only |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.