Learn More
Buerkle™ Mercury Collector
Buerkle™ Mercury Collector enables the easy collection of spilt mercury, which can then be used again.
Brand:  Buerkle™ 9789-0001
47.05 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10483482
  • £52.95 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Enables the easy collection of spilt mercury, which can then be used again
  • The mercury collector is composed of a tub with a screw cap lid that has a special plastic foam inserted
  • On pressing this foam gently on the spilt mercury, the foam's pores are opened and the mercury globules are absorbed
  • The cap is then screwed on the tub, the globules are released again and fall through the perforated separation into the tub
Specifications
Specifications
Mercury Collector
Mercury
Special closure with special plastic foam
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Mercury Collector > Includes: Special closure with special plastic foam