Learn More
Buerkle™ Polyethylene Spigot Jumbo with Tap
Spigot with tap for R2in.barrel thread, 50 mm flow, for viscous media such as lacquers, glue, shampoo.
Brand:  Buerkle™ 0534-0020
Product Code. 10390442
  • £51.55 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cost-effective tap
  • PE, brown
  • Seal made of EPT
Specifications
Specifications
R 2 in. barrel thread
Brown
PE, EPT
Spigot
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Polyethylene Spigot Jumbo with Tap > Color: Brown