Learn More
Uvex Busingess Casual 95388 Insole - Width 11
Comfortable climatic insole with antistatic properties for Uvex 1 business Casual
Brand: Uvex 9538845
Additional Details : Weight : 0.07000kg
Description
- Full surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Perforation for improved breathability
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No. 9538.8
- Size: 38 to 48
Specifications
Black | |
Microvelour | |
No | |
Standard width | |
No |
Uvex Business Casual | |
45 | |
Unisex | |
Yes |