missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butan-1-ol, Certified AR for Analysis, Fisher Chemical™
£64.45 - £894.00
Chemical Identifiers
CAS | 71-36-3 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | 2964 |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
IUPAC Name | butan-1-ol |
SMILES | CCCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10734941
|
Fisher Chemical
B/4850/08 |
500 mL | Amber Glass Bottle |
£64.45
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530911
|
Fisher Chemical
B/4850/15 |
1 L | Amber Glass Bottle |
£92.25
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10141610
|
Fisher Chemical
B/4850/17 |
2.5 L | Amber Glass Bottle |
£145.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10477630
|
Fisher Chemical
B/4850/PB17 |
2.5 L | Plastic-coated Glass Bottle |
£192.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10744941
|
Fisher Chemical
B/4850/25 |
25 L | Metal Can |
£894.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
71-36-3 | |
74.12 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
CCCCO |
C4H10O | |
2964 | |
butan-1-ol |
Specifications
-89°C | |
Colorless | |
Liquid | |
2964 | |
2.95 mPaS ( at 20°C) | |
CCCCO | |
74.12 |
71-36-3 | |
117.6°C | |
C4H10O | |
6.7mbar at 20°C | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
74.12g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Butanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.