missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butan-1-ol, Extra Pure, SLR, Fisher Chemical™
21.62 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
Butan-1-ol, Extra Pure, SLR
Specifications
Melting Point | -89°C |
Color | Colorless |
Boiling Point | 117.6°C |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 6.7mbar at 20°C |
Viscosity | 2.95 mPaS ( at 20°C) |
Formula Weight | 74.12g/mol |
Safety and Handling
Product Identifier
- 1-Butanol
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'