missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benazepril hydrochloride, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support

Specifications
Specifications
CAS | 86541-74-4 |
Packaging | Glass Bottle |
Quantity | 100 mg |
For Use With (Application) | Ace Inhibitors |
Physical Form | Solid |
Grade | Analytical Standard |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Benazepril hydrochloride, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction