missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, Reagent Grade, 99.5%, Honeywell™

Reagent Grade, 99.5%

£31.95 - £104.04

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15655770
View Documents
Honeywell Research Chemicals
537454-1L
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15628420
View Documents
Honeywell Research Chemicals
537454-4L
4 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

123-86-4
116.16
DKPFZGUDAPQIHT-UHFFFAOYSA-N
31272
butyl acetate
C6H12O2
MFCD00009445
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
CHEBI:31328
CCCCOC(C)=O

Specifications

123-86-4
Colorless
126.2°C
23°C (73.4°F)
C6H12O2
n20/D 1.394
UN1123
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
CCCCOC(C)=O
116.16
4 (vs air)
116.16g/mol
99.50%
-78°C
0.88g/mL at 25°C
0.076
790°F
CH3COO(CH2)3CH3
MFCD00009445
1741921
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
31272
CHEBI:31328
15mmHg (25°C), 8mmHg (20°C)
Butyl acetate
Videos
Safety and Handling

Safety and Handling

P210-P370 + P378

missing translation for 'dotInformation' : class 3 - PG 3 - Butyl acetates

missing translation for 'rtecsNumber' : 204-658-1

SDS
Documents

Documents

Product Certifications