missing translation for 'onlineSavingsMsg'
Learn More

Butyl chloroformate, 98%, Thermo Scientific Chemicals

Product Code. 10144150
Click to view available options
Quantity:
5 mL
25 mL
100 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-34-7
MDL Number MFCD00000650
InChI Key NRDQFWXVTPZZAZ-UHFFFAOYSA-N
Synonym butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate
PubChem CID 61140
IUPAC Name butyl carbonochloridate
SMILES CCCCOC(=O)Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H9ClO2
Linear Formula ClCO2(CH2)3CH3
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Quantity 5 mL
Beilstein 03, II, 11
Molecular Weight (g/mol) 136.575
Formula Weight 136.58
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Butyl chloroformate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'