missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl chloroformate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
Specifications
Specifications
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C5H9ClO2 |
Linear Formula | ClCO2(CH2)3CH3 |
Refractive Index | 1.4110 to 1.4130 (20°C, 589nm) |
Quantity | 100 mL |
Beilstein | 03, II, 11 |
Molecular Weight (g/mol) | 136.575 |
Formula Weight | 136.58 |
Show More |
Safety and Handling
Product Identifier
- Butyl chloroformate
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'