missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Digol, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Specifications
Melting Point | -68°C |
Color | Colorless |
Boiling Point | 231°C |
Packaging | Amber glass bottle |
Quantity | 500 mL |
Formula Weight | 162.23g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- 2-(2-Butoxyethoxy)ethanol
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'