missing translation for 'onlineSavingsMsg'
Learn More

Butyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10318640
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-88-1, 150-76-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00009444
InChI Key SOGAXMICEFXMKE-UHFFFAOYSA-N
Synonym butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
PubChem CID 7354
IUPAC Name butyl 2-methylprop-2-enoate
SMILES CCCCOC(=O)C(C)=C

Specifications

Melting Point -75.0°C
Density 0.8900g/mL
Boiling Point 160.0°C to 163.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2(CH2)3CH3
Refractive Index 1.4225 to 1.4245
Quantity 1 L
Beilstein 02, III, 1286
Specific Gravity 0.89
Solubility Information Solubility in water: 3g/L in water (20°C). Other solubilities: very soluble in ether and ethanol,miscible in ethyl acetate
Stabilizer 10 to 110ppm MEHQ
Viscosity 1.1 mPa.s (20°C)
Formula Weight 142.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyl methacrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyl methacrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'