Learn More
Cytiva Butyl-S Sepharose™ 6 Fast Flow Chromatography Media
The least hydrophobic medium in the Sepharose Fast Flow hydrophobic interaction chromatography (HIC) range and is particularly useful during the initial stages of purification process
Brand: Cytiva 17-0978-02
Code : GE
Additional Details : CAS Number : 64-17-5 Weight : 1.33810kg
Description
- Binds and elutes relatively strong hydrophobic molecules at comparatively low salt concentrations
- Hydrophilic nature of the base matrix ensures low levels of non-specific binding
- Purifies recombinant Hepatitis B virus surface antigen from CHO cells
- Composed of cross-linked 6% agarose beads modified with butyl groups via thio-ether linkage
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
200 mL | |
Stable in commonly used aqueous Buffers: 1 mM HCl, 1 M NaOH, 30% Isopropanol, 50% Ethylene Glycol, 70% Ethanol, 6 M Guanidine hydrochloride, 8 M Urea | |
Butyl-S Sepharose 6 fast flow media | |
100 kPa | |
3 to 13 | |
Butyl-S |
For Hydrophobic Medium in The Sepharose Fast Flow Hydrophobic Interaction Chromatography (HIC) Range and is Particularly Useful During The Initial Stages of PurifiCatIon Process | |
4°C to 30°C | |
45 - 165 µm | |
6% Cross-Linked Agarose | |
20% Ethanol |