missing translation for 'onlineSavingsMsg'
Learn More
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals Product Code.: 10063330

Butyric acid sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10063330
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 10063330

Brand: Thermo Scientific Acros 263190050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4H7NaO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00002816
InChI Key MFBOGIVSZKQAPD-UHFFFAOYSA-M
Synonym sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1Show MoreShow Less
PubChem CID 5222465
ChEBI CHEBI:64103
SMILES [Na+].CCCC([O-])=O

Specifications

Melting Point 250.0°C to 253.0°C
Color White
Assay Percent Range 98%
Linear Formula CH3CH2CH2COONa
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether
IUPAC Name sodium;butanoate
Formula Weight 110.09
Percent Purity 99%
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Amorphous Powder
Chemical Name or Material Butyric acid sodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461025 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number263190050,263191000,263190250,263195000
Lot NumberA0451582 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number263190050,263191000,263190250,263195000
Lot NumberA0449625 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog Number263190050,263191000,263190250,263195000
Lot NumberA0393786 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number263190050,263191000,263190250,263195000
Lot NumberA0410372 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number263190050,263191000,263190250,263195000
compliance-icons
Product Identifier
  • Butyric acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.