missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyroin, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Boiling Point | 80.0°C to 86.0°C (12.0 mmHg) |
Flash Point | 71°C |
Assay Percent Range | 96% min. (GC) |
Quantity | 5 g |
Formula Weight | 144.22 |
Percent Purity | 97% |
Chemical Name or Material | Butyroin |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'