missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BUV395 Streptavidin
Affinity purified
Brand: BD 564176
Additional Details : Weight : 0.01000kg
Description
Host Species:
Species Reactivity: Mouse
Flow Cytometry
Specifications
Streptavidin | |
Mouse | |
Affinity-purified |
BUV395 | |
100 μg |