missing translation for 'onlineSavingsMsg'
Learn More

BW Technologies™ GasAlertQuattro Multi Gas Detector

Rugged and reliable four-gas detector combines a comprehensive range of features with simple one-button operation. BW Technologies™ GasAlertQuattro Multi Gas Detector features flexible power options that keep it always ready to use.

Brand:  BW Technologies™ QT-XWHM-R-Y-E-00

Product Code. 11978995

  • £1525.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Minimize costs and training with one-button operation.
  • Field-proven Surecell sensors offer an unprecedented performance in even the harshest environments.
  • Interchangeable power options with extended battery runtimes for longer shifts.
  • Continuous LCD shows real-time gas concentrations.
  • Compact and lightweight design makes it comfortable to wear.
  • Simple automatic calibration procedure; compatible with BW MicroDock II automatic test and calibration station.
  • Full function self-test of sensor(s), battery status, circuit integrity and audible/visual alarms on start up and continuous testing on sensor(s).
  • Bright wide-angled visual alarm bars.
  • Built-in concussion-proof boot.
  • Powered by an interchangeable rechargeable battery pack or alkaline pack with 3 AA batteries.
  • One-button operation and straightforward user interface minimizes training.
  • Comprehensive datalogging and event logging capacity.
  • IntelliFlash verifies operation and compliance to both the user and supervisors from up to 20ft. / 6.1m.
  • Enhanced resistance to common industrial cross sensitive gases such as methanol and ethanol (CO and H2S sensors).
  • Multi-language support in English, French, German, Spanish and Portuguese.
  • Field-proven Surecell sensors offer an unprecedented performance in even the harshest environments.
  • 4 channel monitor with 4 sensors.
  • SureCell H2S and CO sensors are superior in very cold/dry and hot/humid conditions.
  • SureCell is a patented and proven Honeywell sensor in use for several years.
  • Run time on rechargeable batteries is 21 hours at 70 degrees F.
  • Alkaline batteries can also be used.
  • Larger LCD screen and graphics.
  • One button, easy to use.
  • Intellisense green flashing light indicates unit is on, calibrated and bump tested.
  • Reflex Technology monitor continuously self checks sensors, battery, circuit, memory.
Specifications

Specifications

Audible
Lithium, Rechargeable
47 mm
Gas Alert Dectector
81 mm
1 to 0.1 %
316g
20 hr.
Ex II 1G Ga Ex ia IIC T4
Oxygen, Carbon Monoxide, Hydrogen Sulfide, LEL
130 mm
0 to 100%
24Months
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.