missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BX 471, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective non-peptide CCR1 antagonist (Ki = 1 nM for human CCR1). Exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4. Inhibits MIP-α/CCL3-induced intracellular Ca2+ mobilization. Orally active; effectively reduces disease severity in a rat model of multiple sclerosis. Decreases renal fibrosis in a mouse model of obstructive nephropathy.
Specifications
Quantity | 10 mg |
Formula Weight | 434.89 |
Percent Purity | >98% |
Chemical Name or Material | BX 471 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'