missing translation for 'onlineSavingsMsg'
Learn More

BX 513 hydrochloride, Tocris Bioscience™

Product Code. 18014082
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective CCR1 antagonist

Selective CCR1 receptor antagonist (Ki values are 0.04, > 10, > 10 and > 10 nM for CCR1, CCR5, CXCR2 and CXCR4 receptors respectively). Inhibits MIP-1α-induced intracellular calcium mobilization (IC50 = 2.5 μM). Also a full inverse agonist at US28, a HCMV-encoded chemokine receptor.

Chemical Identifiers

CAS 1216540-18-9
Molecular Formula C28H30Cl2N2O
Molecular Weight (g/mol) 481.461
InChI Key SSZWNUGWOGONQJ-UHFFFAOYSA-N
Synonym bx 513 hydrochloride, bx-513 hydrochloride, 1-piperidinepentanenitrile,4-4-chlorophenyl-4-hydroxy-a,a-diphenyl-,hydrochloride, 4-4-chlorophenyl-4-hydroxy-?,?-diphenyl-1-piperidinepentanenitrile hydrochloride, 5-4-4-chlorophenyl-4-hydroxypiperidin-1-yl-2,2-diphenylpentanenitrile hydrochloride, 5-4-4-chlorophenyl-4-hydroxypiperidin-1-yl-2,2-diphenylpentanenitrile-hydrogen chloride 1/1
PubChem CID 56972186
IUPAC Name 5-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-2,2-diphenylpentanenitrile;hydrochloride
SMILES C1CN(CCC1(C2=CC=C(C=C2)Cl)O)CCCC(C#N)(C3=CC=CC=C3)C4=CC=CC=C4.Cl

Specifications

Quantity 10 mg
Formula Weight 481.46
Percent Purity >99%
Chemical Name or Material BX 513 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.