missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Cbl Monoclonal Antibody (S.551.7), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514955
254.01 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Cbl-b or Cbl-c proteins.
Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
Specifications
c-Cbl | |
Monoclonal | |
18.6 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P22681 | |
Cbl | |
Synthetic peptide corresponding to the sequence of human c-Cbl | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
S.551.7 | |
Unconjugated | |
Cbl | |
4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL | |
Rabbit | |
Affinity chromatography | |
RUO | |
867 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
c-Cbl Monoclonal Antibody (S.551.7), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction