missing translation for 'onlineSavingsMsg'
Learn More

Cytiva C Columns

C columns are economical modular columns for low pressure or gravity feed, providing even distribution of eluent to the medium bed, even without flow adapters.

Brand:  Cytiva 19-5202-01

 View more versions of this product

Product Code. 10556900

  • £358.00 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Columns are available in three diameters and several lengths.
  • Compatible with a variety of bulk media.
  • Flow adapters are available for each of the column diameters to allow optimal bed heights for each application, as are thermostatic jackets for precise temperature control.
  • Packing reservoirs for each column diameter optimize packing conditions.
  • C columns are delivered with polyethylene tubing (1.0-mm ID, 2 m in length), bed support screens, snap-on net rings, and instructions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C Chromatography Columns
700mm
Borosilicate glass
Polyamide/Polypropylene net support
Low pressure or gravity fed separations
26mm
Polyethylene tubing (1.0mm I.D., 2m length), bed support screens, snap-on net rings and instructions
14psi (1 bar)
Empty Chromatography Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers