Learn More
Tocris Bioscience™ c-Di-GMP sodium salt
Endogenous STING and DDX41 agonist; activates STING-dependent signaling
Brand: Tocris Bioscience™ 5900/730U
Additional Details : Weight : 0.00970kg
Description
c-Di-GMP sodium salt is an endogenous STING and DDX41 agonist; mediates DDX41-STING interaction. Activates STING-dependent IFN-β production in mouse and human cells. Also induces an antitumor response in a lung B16-10 melanoma mouse model, when administered intravenously within YSKO5-liposomes.This product is sold in units of 730μg, equivalent to 1μmol.
Specifications
3′,5′-Cyclic diguanylic acid sodium salt | |
730 μg | |
C20H22N10Na2O14P2 | |
>98% |
endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membr | |
STING/TMEM173 | |
734.37 |