missing translation for 'onlineSavingsMsg'
Learn More

3M Indicator tape Comply ethylene oxide 19mm x 55m

Brand:  3M 1224-6

Indicator tape Comply ethylene oxide 19mm x 55m

Code : 04

Additional Details : Weight : 0.01000kg

Product Code. 12318209

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers