missing translation for 'onlineSavingsMsg'
Learn More

Merck Sample preparation device Labscale TFF System 230V 50Hz has a graduated 500mL acrylic reservoir

Brand:  Merck XX42LSS13

Sample preparation device Labscale TFF System 230V 50Hz has a graduated 500mL acrylic reservoir

Code : 72

Additional Details : Weight : 9.50000kg

Product Code. 12811676

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers