missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf58 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20446-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in redox regulation of the cell. Acts as an antioxidant. Inhibits TNFSF11-induced NFKB1 and JUN activation and osteoclast differentiation. May affect bone resorption and help to maintain bone mass.Specifications
C10orf58 | |
Polyclonal | |
Unconjugated | |
FAM213A | |
C10orf58, F213A, FAM213A, PSEC0139, UNQ611/PRO1198, UPF0765 protein C10orf58 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361118, 84293 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6AXX6, Q9BRX8 | |
Fam213a | |
C10orf58 Fusion Protein Ag14272 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction