missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | C16orf70 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810165
|
Proteintech
20602-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800165
|
Proteintech
20602-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MW of this protein is 48 kDa, and our antibody specifically recognizes the 48 kDa protein.
The function of C20orf30 has not been widely studied, and is yet to be fully elucidated.Specifications
C16orf70 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q9BSU1 | |
102547323, 80262 | |
C16orf70 Fusion Protein Ag14576 | |
Primary | |
-20°C | |
C16orf70 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C16orf6, C16orf70, lin 10, LIN10, UPF0183 protein C16orf70 | |
C16orf70, LOC102547323 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |