missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C21orf58 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25024-1-AP-20UL
This item is not returnable.
View return policy
Specifications
C21orf58 | |
Polyclonal | |
Unconjugated | |
C21orf58 | |
C21orf58 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54058 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P58505 | |
C21orf58 | |
C21orf58 Fusion Protein Ag21758 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |