missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf47 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24930-1-AP-150UL
This item is not returnable.
View return policy
Description
Promotes sorting of SMDT1/EMRE in mitochondria by ensuring its maturation. Interacts with the transit peptide region of SMDT1/EMRE precursor protein in the mitochondrial matrix, leading to protect it against protein degradation by YME1L1, thereby ensuring SMDT1/EMRE maturation by the mitochondrial processing peptidase (PMPCA and PMPCB).Specifications
C2orf47 | |
Polyclonal | |
Unconjugated | |
MAIP1 | |
C2orf47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68115, 79568 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BHE8, Q8WWC4 | |
MAIP1 | |
C2orf47 Fusion Protein Ag21518 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |