missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf43 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | C3orf43 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863945
|
Proteintech
25362-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853945
|
Proteintech
25362-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMCO1 is a protein coding gene.Specifications
C3orf43 | |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q147U7, Q8CEZ1 | |
255798, 69576 | |
C3orf43 Fusion Protein Ag21670 | |
Primary | |
-20°C | |
SMCO1 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C3orf43 | |
Smco1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |