missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf211 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25124-1-AP-150UL
This item is not returnable.
View return policy
Description
ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.Specifications
C6orf211 | |
Polyclonal | |
Unconjugated | |
ARMT1 | |
C6orf211, UPF0364 protein C6orf211 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79624 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H993 | |
ARMT1 | |
C6orf211 Fusion Protein Ag19090 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |