Learn More
CA052 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557510
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PEEPPKEFKI WKSNYVPPPE TYTTEKKPPP PELDMAIKWS NIYEDNGDDA PQNAKKARLL PEGEETLESD DEKDEHTSKK RKVEPGEPAK KK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
C1orf52 gene ontology annotations related to this gene include RNA binding.Specifications
CA052 | |
Polyclonal | |
Unconjugated | |
C1orf52 | |
BAG; BCL10-associated gene protein; C1orf52; chromosome 1 open reading frame 52; gm117; RP11-234D19.1; UPF0690 protein C1orf52 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148423 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6N3 | |
C1orf52 | |
Recombinant protein corresponding to Human CA052 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only