missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CA19-9 Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHCA199X5

 View more versions of this product

Product Code. 17165369

  • £2320.00 / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Human CA19-9 ELISA quantitates Hu CA19-9 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CA19-9. Principle of the method The Human CA19-9 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Mucin glucoprotein is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.274 to 200 U/mL
0.3 U/mL to 200 U/mL
Biotin
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF-7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP- 2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
<10%
HRP
RUO
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L
Human
4 hr. 45 min.
0.3 U/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
<12%
Pre-coated 96 well plates, Biotinylated Detection Antibody, Assay Diluent Concentrate, Stop Solution, Adhesive Plate Covers, Standard, SAV-HRP, Wash Buffer, Chromogen
5 x 96 Tests
Other Research
2°C to 8°C
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers