missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Cabinet for Acids and Alkalis SL-CLASSIC

Safe and approved storage of aggressive, non flammable hazardous materials in working areas

Brand:  asecos™ 3061300130614

1784.74 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 15589823

  • £2165.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For indoor use only

This item is not returnable. View return policy

Description

Description

  • Robust construction and longevity: cabinet body made of powder-coated sheet steel, safety elements assembled outside the storage compartment for increased protection against corrosion
  • Two in one: two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • ventilation: integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
  • Certificates
    • Type tested in accordance with EN 14727
    • EN 16121/16122 conformity
    • GS certified
Specifications

Specifications

DIN EN 16121/16122 conformity, GS certified
603 mm
Cabinet for acids and alkalis SL-CLASSIC
603 x 597 x 1965 mm
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
914 mm
2
Pull-out
166.60 kg
485 mm
Chipboard
4 x pull-out shelf (panel melamine resin-coated/PP)
570 mm
Top: 570 x 485 x 914 mm, Bottom: 570 x 485 x 833 mm
Wing, Right-hinged
1965 mm
SL.196.060.MH.R
4
Melamine resin laminated
597 mm
Light Gray
Cabinet for acids and alkalis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.