missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
£545.00 - £600.00
Specifications
Product Type | Soft, Flexible Animal Feeding Needle |
---|---|
Material | PTFE |
Disposable | Yes |
Sterility | Sterile |
Product Code | Brand | Diameter (English) Ball | Gauge | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Ball | Gauge | Length (English) | Price | Quantity & Availability | |||
17614825
|
Cadence Science™
9924 |
0.078 in. | 18G | 3 in. |
£600.00
Pack of 100 |
Estimated Shipment: 31-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15786118
|
Cadence Science™
9931 |
0.074 in. | 20G | 1.2 in. |
£600.00
Pack of 100 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15282065
|
Cadence Science™
9928 |
0.078 in. | 18G | 1.5 in. |
£545.00
Pack of 100 |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Nontoxic, nonpyrogenic
- PTFE tubing is flexible and softer than other types of tubing
- Low friction protects delicate organs
- Translucent for easy confirmation of reagent delivery
- Plastic luer hubs; soft silicone tips
- Sterile, packaged in pouches
- Five needles per pouch, 20 pouches per box
Specifications
Soft, Flexible Animal Feeding Needle | |
PTFE | |
Yes | |
Sterile |