Learn More
Cadmium acetate dihydrate, 98%, pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 250 g |
Assay Percent Range | 97.5% min. (Complexometry) |
Linear Formula | (CH3CO2)2Cd·2H2O |
Merck Index | 10,1584 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol |
Formula Weight | 266.52 |
Percent Purity | 98% |
Grade | Pure |
Loss on Drying | 12.5% to 14.5% |
Chemical Name or Material | Cadmium acetate dihydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464278 | Certificate of Analysis | 16/07/2024 | 212310050,212312500,212310010 |
A0459429 | Certificate of Analysis | 22/01/2024 | 212310050,212312500,212310010 |
A0430792 | Certificate of Analysis | 30/11/2023 | 212310050,212312500,212310010 |
A0438183 | Certificate of Analysis | 16/08/2023 | 212310050,212312500,212310010 |
A0402582 | Certificate of Analysis | 15/12/2022 | 212310050,212312500,212310010 |
Safety and Handling



- Cadmium acetate dihydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.