missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate dihydrate, 98%

Product Code. 11358698
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11358698

Brand: Thermo Scientific Alfa Aesar 041581.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In cadium electroplating | CAS: 5743-04-4 | C4H10CdO6 | 266.53 g/mol

Cadmium acetate dihydrate is in the porcelain production for the production of iridescent used effects. Moreover, it is in the analytical chemistry, a reagent for the detection of sulfur, selenium and tellurium. It is also used as chemical and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00012449
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Color White
Physical Form Powder
Quantity 500 g
UN Number UN2570
Merck Index 14,1614
Solubility Information Freely soluble in water, soluble in alcohol
Formula Weight 266.54 (230.50 Anhydrous)
Percent Purity 98%
Sensitivity Hygroscopic
Density 2.01 g/mL
Chemical Name or Material Cadmium acetate dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.