missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Honeywell™

Product Code. 15651880 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 15651880

Brand: Honeywell Research Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Melting Point 256°C
Physical Form Solid
Quantity 500 g
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity ≥98.0% (KT)
Grade purum p.a.
Packaging Poly Bottle
Density 2.01g/cu.cm
Chemical Name or Material Cadmium Acetate Dihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cadmium acetate dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Cadmium acetate-2- hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.