missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide, 98%, pure, anhydrous, Thermo Scientific Chemicals

£32.66 - £286.43

Chemical Identifiers

CAS 7789-42-6
Molecular Formula Br2Cd
Molecular Weight (g/mol) 272.22
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15289886
View Documents
Thermo Scientific Acros
453860250
25 g
£32.66
25g
Please to purchase this item. Need a web account? Register with us today!
15299886
missing translation for 'viewDocuments'
Thermo Scientific Acros
453861000
100 g
£96.78
100g
Please to purchase this item. Need a web account? Register with us today!
15209896
missing translation for 'viewDocuments'
Thermo Scientific Acros
453865000
500 g
£286.43
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-42-6
Br2Cd
272.22
cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

963.0°C
7789-42-6
Br2Cd
cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
272.22
98%
Cadmium bromide
566.0°C
97.5% min
15,162
Solubility in water: soluble. Other solubilities: soluble in alcohols, moderatly soluble in acetone, slightly soluble in ether
272.22
Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 232-165-1

Product Identifier
  • Cadmium bromide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only