missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11455210
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11455210

Brand: Thermo Scientific Alfa Aesar 044498.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681435-25-6
Molecular Weight (g/mol) 272.22
MDL Number MFCD00150204
InChI Key KPWJBEFBFLRCLH-UHFFFAOYSA-L
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
IUPAC Name cadmium(2+) dibromide
SMILES [Br-].[Br-].[Cd++]

Specifications

Color White to Yellow
Physical Form Crystalline
Quantity 100 g
Molecular Formula Br2Cd
UN Number UN2570
Formula Weight 272.22 (Anhydrous)
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.