missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ACS reagent, anhydrous

Product Code. 10682145 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10682145

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960.0°C
Melting Point 568.0°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula CdCl2
Fieser 04,485
Merck Index 15,1622
Solubility Information Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
Formula Weight 183.31
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium chloride, Anhydrous, For analysis ACS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : cadmium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.