missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Chloride, Honeywell™

Puriss. p.a., anhydrous,≥99.0%

£51.20 - £455.99

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15611880
View Documents
Honeywell Research Chemicals
20899-25G
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15601880
View Documents
Honeywell Research Chemicals
20899-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15621880
View Documents
Honeywell Research Chemicals
20899-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

10108-64-2
183.31
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
CHEBI:35456
[Cl-].[Cl-].[Cd++]

Specifications

10108-64-2
CdCl2
2570
Soluble in water 457g/L at 20°C
[Cl-].[Cl-].[Cd++]
183.31
CHEBI:35456
≥99%
10mmHg (656°C)
Cadmium Chloride
Anhydrous
MFCD00010916
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.32g/mol
puriss. p.a.
Vapor Density: 6.3 (vs air)
Videos
Safety and Handling

Safety and Handling

P201-P260-P280-P304 + P340 + P310-P308 + P313

H301-H330-H340-H350-H360FD-H372-H410

missing translation for 'einecsNumber' : 233-296-7

SDS
Documents

Documents

Product Certifications