missing translation for 'onlineSavingsMsg'
Learn More
Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11380879

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11380879
50 g
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11380879

Brand: Thermo Scientific Alfa Aesar 010661.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654054-66-7
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00149627
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
PubChem CID 16211517
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Physical Form Crystalline Powder
Quantity 50 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water and acetone. Slightly soluble in methanol. Insoluble in ether.
IUPAC Name dichlorocadmium;hydrate
Formula Weight 183.32 (Anhydrous)
Percent Purity 99.998%
Grade Puratronic™
Refractive Index 1.6513
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.