missing translation for 'onlineSavingsMsg'
Learn More

Cadmium foil, 0.025mm (0.001 in.) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

£85.02 - £1061.00

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow MoreShow Less
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11310649
SDS View Documents
Thermo Scientific Alfa Aesar
000064.FQ
25 x 100 mm
£85.02
Each
Please to purchase this item. Need a web account? Register with us today!
11320649
SDS View Documents
Thermo Scientific Alfa Aesar
000064.FV
25 x 400 mm
£255.27
Each
Please to purchase this item. Need a web account? Register with us today!
11300649
SDS View Documents
Thermo Scientific Alfa Aesar
000064.HA
25 x 2000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium foil, 0.025mm (0.001 in.) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.