missing translation for 'onlineSavingsMsg'
Learn More

Cadmium foil, 0.05mm (0.002in) thick, Puratronic™, 99.999% (metals basis)

Product Code. 15426585
Click to view available options
Quantity:
50 x 50 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15426585

Brand: Thermo Scientific Alfa Aesar 044127.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-43-9 | Cd | 112.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-43-9
Quantity 50 x 50 mm
Weight ≈1.1g/50x50mm
Form Foil
Molecular Formula Cd
MDL Number MFCD00010914
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES [Cd]
IUPAC Name cadmium
Molecular Weight (g/mol) 112.41
PubChem CID 23973
ChEBI CHEBI:22977
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Cadmium foil, 0.05mm (0.002 in.) thick
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.